siRNA Details
| Gene Symbol: Homer2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164086.1 | NP_001157558.1 | 2 | 292 | ||
| NM_001164087.1 | NP_001157559.1 | 2 | 275 | |||
| NM_011983.2 | NP_036113.1 | 2 | 292 | |||
| GenBank mRNA | AB017136.1 | - | 2 | 318 | ||
| AF093259.1 | - | 2 | 197 | |||
| AF093260.1 | - | 2 | 197 | |||
| AJ012076.1 | - | 2 | 144 | |||
| AK158673.1 | - | 234 | ||||
| AK163423.1 | - | 2 | 293 | |||
| CJ083069.1 | - | 2 | 276 | |||
| DV657327.1 | - | 2 | 197 | |||

Homer2
Genomic Map