siRNA Details
| Gene Symbol: Homer2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164086.1 | NP_001157558.1 | 9 | 1076 | ||
| NM_001164087.1 | NP_001157559.1 | 9 | 1092 | |||
| NM_011983.2 | NP_036113.1 | 9 | 1109 | |||
| GenBank mRNA | AB017136.1 | - | 9 | 1102 | ||
| AF093259.1 | - | 9 | 981 | |||
| AF093260.1 | - | 9 | 1014 | |||
| AK003632.1 | - | 1 | 22 | |||
| AK158673.1 | - | 1178 | ||||
| BC038314.1 | - | 5 | 750 | |||

Homer2
Genomic Map