siRNA Details
| Gene Symbol: TTC27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193509.1 | NP_001180438.1 | 2 | 253 | ||
| NM_017735.4 | NP_060205.3 | 2 | 500 | |||
| XM_005264416.1 | XP_005264473.1 | 2 | 487 | |||
| XM_011532958.1 | XP_011531260.1 | 2 | 487 | |||
| GenBank mRNA | AK027570.1 | - | 2 | 409 | ||
| AK299196.1 | - | 2 | 242 | |||
| BC001248.1 | - | 1 | 33 | |||
| BC063791.1 | - | 2 | 458 | 6500951 | ||
| BI546117.1 | - | 2 | 518 | |||

TTC27
Genomic Map