siRNA Details
| Gene Symbol: Magee2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053206.2 | NP_444436.1 | 1 | 504 | ||
| GenBank mRNA | AK036343.1 | - | 1 | 505 | ||
| BC094361.1 | - | 1 | 393 | 6829915 | ||
| BC132640.1 | - | 1 | 359 | 40130917 | ||
| BC138210.1 | - | 1 | 359 | 8861229 | ||

Magee2
Genomic Map