siRNA Details
| Gene Symbol: CWC25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017748.4 | NP_060218.1 | 3 | 491 | ||
| NR_073428.1 | 4 | 608 | ||||
| GenBank mRNA | AK000298.1 | - | 3 | 441 | ||
| AK225373.1 | - | 3 | 462 | |||
| AK296115.1 | - | 4 | 573 | |||
| AK302983.1 | - | 3 | 392 | |||
| AK316077.1 | - | 2 | 245 | |||
| BC003085.1 | - | 3 | 344 | |||
| BC008833.2 | - | 3 | 477 | 4304656 | ||
| HY021361.1 | - | 3 | 492 | |||

CWC25
Genomic Map