siRNA Details
| Gene Symbol: Olr652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000638.1 | NP_001000638.1 | 571 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr652| Gene Symbol: Olr652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000638.1 | NP_001000638.1 | 571 | |||