siRNA Details
| Gene Symbol: KCTD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017634.3 | NP_060104.2 | 11, 12 | 1269 | ||
| GenBank mRNA | AK000045.1 | - | 6, 7 | 597 | ||
| AK298518.1 | - | 10, 11 | 867 | |||
| BC021216.2 | - | 11, 12 | 1072 | 4302317 | ||
| BC068518.1 | - | 11, 12 | 1082 | 5265435 | ||

KCTD9
Genomic Map