siRNA Details
| Gene Symbol: Olr1253 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000805.1 | NP_001000805.1 | 1 | 913 | ||
| Gene Symbol: LOC100912543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762453.1 | XP_008760675.1 | 1 | 913 | ||
| Gene Symbol: LOC103692041 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762675.1 | XP_008760897.1 | 1 | 913 | ||

Olr1253
Genomic Map