siRNA Details
| Gene Symbol: Olr545 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001053.1 | NP_001001053.1 | 835 | |||
| Gene Symbol: LOC103691842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762001.1 | XP_008760223.1 | 1 | 850 | ||

Olr545
Genomic Map