siRNA Details
| Gene Symbol: C12orf29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009894.2 | NP_001009894.2 | 7 | 2663 | ||
| GenBank mRNA | AK095477.1 | - | 7 | 2663 | ||
| AL049382.1 | - | 1 | 1389 | |||
| BC035136.2 | - | 6 | 2500 | |||
| BC092422.1 | - | 7 | 2605 | 30386540 | ||
| BX537868.1 | - | 7 | 2635 | |||
| BX640968.1 | - | 7 | 2638 | |||
| BX648026.1 | - | 6 | 2499 | |||
| CR936776.1 | - | 7 | 2653 | |||

C12orf29
Genomic Map