siRNA Details
| Gene Symbol: MYBPHL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010985.2 | NP_001010985.2 | 8 | 1119 | ||
| NM_001265613.1 | NP_001252542.1 | 8 | 1050 | |||
| XM_017001173.1 | XP_016856662.1 | 8 | 3472 | |||
| XM_017001174.1 | XP_016856663.1 | 8 | 3472 | |||
| XM_017001175.1 | XP_016856664.1 | 8 | 3403 | |||
| GenBank mRNA | AK129834.1 | - | 8 | 1119 | ||
| BC108664.1 | - | 3 | 352 | |||
| BC132900.1 | - | 8 | 1096 | 40146922 | ||
| BC132902.1 | - | 8 | 1096 | 40146924 | ||
| BC144471.1 | - | 8 | 1027 | 9052998 | ||

MYBPHL
Genomic Map