siRNA Details
| Gene Symbol: Inpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012131.1 | NP_001012131.1 | 4 | 685 | ||
| XM_006244899.2 | XP_006244961.1 | 5 | 905 | |||
| XM_006244901.2 | XP_006244963.1 | 4 | 704 | |||
| XM_008767091.1 | XP_008765313.1 | 5 | 1646 | |||
| XM_008767092.1 | XP_008765314.1 | 5 | 757 | |||
| XM_008767093.1 | XP_008765315.1 | 3 | 2797 | |||
| GenBank mRNA | BC086600.1 | - | 4 | 685 | 7314378 | |

Inpp1
Genomic Map