siRNA Details
| Gene Symbol: Mlph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012135.1 | NP_001012135.1 | 6 | 742 | ||
| XM_006245404.2 | XP_006245466.1 | 6 | 943 | |||
| XM_006245405.2 | XP_006245467.1 | 6 | 945 | |||
| GenBank mRNA | BC081894.1 | - | 6 | 742 | 7110013 | |

Mlph
Genomic Map