siRNA Details
| Gene Symbol: Fus | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012137.1 | NP_001012137.1 | 12 | 1327 | ||
| XM_006230290.2 | XP_006230352.1 | 12 | 1351 | |||
| GenBank mRNA | BC087153.1 | - | 12 | 1327 | 7132489 | |
| FQ220042.1 | - | 4 | 436 | |||

Fus
Genomic Map