siRNA Details
| Gene Symbol: ARSI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012301.2 | NP_001012301.1 | 2 | 1118 | ||
| GenBank mRNA | AB448735.1 | - | 2 | 1109 | ||
| AK122641.1 | - | 2 | 262 | |||
| AY875937.1 | - | 2 | 529 | |||
| BC129995.1 | - | 2 | 586 | 40127932 | ||
| BC129996.1 | - | 2 | 586 | 40127934 | ||
| DA264664.1 | - | 2 | 248 | |||

ARSI
Genomic Map