siRNA Details
| Gene Symbol: ANO9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012302.2 | NP_001012302.2 | 18 | 1700 | ||
| XM_011520053.2 | XP_011518355.1 | 18 | 1796 | |||
| XM_017017644.1 | XP_016873133.1 | 19 | 1830 | |||
| XM_017017645.1 | XP_016873134.1 | 19 | 1782 | |||
| XM_017017646.1 | XP_016873135.1 | 18 | 1719 | |||
| XM_017017647.1 | XP_016873136.1 | 18 | 1671 | |||
| GenBank mRNA | AK026923.1 | - | 18 | 1647 | ||
| AK074088.1 | - | 13 | 3482 | |||
| AK096874.1 | - | 9 | 988 | |||
| AL832289.1 | - | 8 | 889 | |||
| BC128557.1 | - | 18 | 1668 | 40122929 | ||
| BC144415.1 | - | 17 | 1726 | |||

ANO9
Genomic Map