siRNA Details
| Gene Symbol: Pgbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012311.2 | NP_001012311.2 | 4 | 2723 | ||
| NM_001162919.1 | NP_001156391.1 | 5 | 2892 | |||
| NM_001162920.1 | NP_001156392.1 | 5 | 2834 | |||
| GenBank mRNA | BC089360.1 | - | 4 | 2571 | 30613267 | |

Pgbd1
Genomic Map