siRNA Details
| Gene Symbol: Nono | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012356.1 | NP_001012356.1 | 12 | 1628 | ||
| GenBank mRNA | BC089880.1 | - | 12 | 1628 | 7366190 | |
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_023144, NM_001252518, AK169303, AK165855, S64860, and others.

Nono
Genomic Map