siRNA Details
| Gene Symbol: ZNF260 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012756.2 | NP_001012774.1 | 4 | 2663 | ||
| NM_001166036.1 | NP_001159508.1 | 4 | 2657 | |||
| NM_001166037.1 | NP_001159509.1 | 3 | 2549 | |||
| NM_001166038.1 | NP_001159510.1 | 3 | 2530 | |||
| XM_011526897.2 | XP_011525199.1 | 2 | 2472 | |||
| XM_017026741.1 | XP_016882230.1 | 2 | 2977 | |||
| GenBank mRNA | AK122854.1 | - | 1 | 1521 | ||
| AK123830.1 | - | 1 | 1185 | |||
| BC042676.1 | - | 4 | 2585 | 4821139 | ||
| BC119003.1 | - | 3 | 2455 | 40082016 | ||
| BC122549.1 | - | 2 | 2328 | 40082014 | ||

ZNF260
Genomic Map