siRNA Details
| Gene Symbol: PABPC1L2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042506.1 | NP_001035971.1 | 1 | 1559 | ||
| GenBank mRNA | BC041956.1 | - | 1 | 1559 | ||
| Gene Symbol: PABPC1L2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012977.2 | NP_001012995.1 | 1 | 1555 | ||

PABPC1L2B
Genomic Map