siRNA Details
| Gene Symbol: TCEAL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012979.2 | NP_001012997.1 | 3 | 961 | ||
| GenBank mRNA | AF086094.1 | - | 1 | 63 | ||
| BC132922.1 | - | 3 | 838 | 40146944 | ||
| BC132924.1 | - | 3 | 838 | 40146946 | ||

TCEAL5
Genomic Map