siRNA Details
| Gene Symbol: TCEAL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012979.2 | NP_001012997.1 | 3 | 891 | ||
| GenBank mRNA | BC132922.1 | - | 3 | 768 | 40146944 | |
| BC132924.1 | - | 3 | 768 | 40146946 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TCEAL5| Gene Symbol: TCEAL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012979.2 | NP_001012997.1 | 3 | 891 | ||
| GenBank mRNA | BC132922.1 | - | 3 | 768 | 40146944 | |
| BC132924.1 | - | 3 | 768 | 40146946 | ||