siRNA Details
| Gene Symbol: SATL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012980.2 | NP_001012998.2 | 1 | 192 | ||
| XM_011530940.2 | XP_011529242.1 | 2 | 626 | |||
| GenBank mRNA | BC043215.1 | - | 1 | 393 | 5295273 | |
| BC126401.1 | - | 1 | 378 | 8992117 | ||

SATL1
Genomic Map