siRNA Details
| Gene Symbol: Hes6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013179.1 | NP_001013197.1 | 3 | 773 | ||
| XM_006245407.2 | XP_006245469.1 | 4 | 726 | |||
| GenBank mRNA | BC087597.1 | - | 3 | 773 | 7311379 | |
| FQ212129.1 | - | 1 | 117 | |||

Hes6
Genomic Map