siRNA Details
| Gene Symbol: PRAMEF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013407.2 | NP_001013425.2 | 4 | 1805 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRAMEF5| Gene Symbol: PRAMEF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013407.2 | NP_001013425.2 | 4 | 1805 | ||