siRNA Details
| Gene Symbol: Trim6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013616.2 | NP_001013637.1 | 8 | 2134 | ||
| GenBank mRNA | AK033396.1 | - | 8 | 2114 | ||
| AK049238.1 | - | 8 | 2114 | |||
| BC026912.1 | - | 1 | 1358 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim6| Gene Symbol: Trim6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013616.2 | NP_001013637.1 | 8 | 2134 | ||
| GenBank mRNA | AK033396.1 | - | 8 | 2114 | ||
| AK049238.1 | - | 8 | 2114 | |||
| BC026912.1 | - | 1 | 1358 | |||