siRNA Details
| Gene Symbol: IGLL3P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029395.1 | 3 | 643 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IGLL3P| Gene Symbol: IGLL3P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029395.1 | 3 | 643 | |||