siRNA Details
| Gene Symbol: TMEM236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098844.2 | NP_001092314.1 | 4 | 3540 | ||
| GenBank mRNA | AK056775.1 | - | 1 | 1099 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM236| Gene Symbol: TMEM236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098844.2 | NP_001092314.1 | 4 | 3540 | ||
| GenBank mRNA | AK056775.1 | - | 1 | 1099 | ||