siRNA Details
| Gene Symbol: AADACL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013630.1 | NP_001013652.1 | 4 | 1448 | ||
| XM_017001152.1 | XP_016856641.1 | 4 | 1373 | |||
| XM_017001153.1 | XP_016856642.1 | 2 | 1410 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AADACL4| Gene Symbol: AADACL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013630.1 | NP_001013652.1 | 4 | 1448 | ||
| XM_017001152.1 | XP_016856641.1 | 4 | 1373 | |||
| XM_017001153.1 | XP_016856642.1 | 2 | 1410 | |||