siRNA Details
| Gene Symbol: Apool | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014105.1 | NP_001014127.1 | 5, 6 | 537 | ||
| XM_006257191.2 | XP_006257253.1 | 5, 6 | 476 | |||
| GenBank mRNA | BC086434.1 | - | 5, 6 | 537 | 7302620 | |
| FQ219134.1 | - | 5, 6 | 427 | |||
| FQ235331.1 | - | 5, 6 | 423 | |||

Apool
Genomic Map