siRNA Details
| Gene Symbol: Mospd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014107.1 | NP_001014129.1 | 7 | 1914 | ||
| XM_006257633.2 | XP_006257695.1 | 6 | 1957 | |||
| XM_006257634.2 | XP_006257696.1 | 6 | 1848 | |||
| GenBank mRNA | BC086521.1 | - | 7 | 1914 | 7302680 | |
| Gene Symbol: LOC686087 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006251400.2 | XP_006251462.1 | 2 | 1930 | ||
| XM_008770405.1 | XP_008768627.1 | 3 | 1821 | |||

Mospd1
Genomic Map