siRNA Details
| Gene Symbol: Gpc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014108.1 | NP_001014130.1 | 9 | 2604 | ||
| XM_006257580.2 | XP_006257642.1 | 10 | 2202 | |||
| GenBank mRNA | BC081962.1 | - | 9 | 2604 | 7124728 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpc4| Gene Symbol: Gpc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014108.1 | NP_001014130.1 | 9 | 2604 | ||
| XM_006257580.2 | XP_006257642.1 | 10 | 2202 | |||
| GenBank mRNA | BC081962.1 | - | 9 | 2604 | 7124728 | |