siRNA Details
| Gene Symbol: Msl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014111.1 | NP_001014133.1 | 5 | 513 | ||
| XM_006256835.2 | XP_006256897.2 | 6 | 899 | |||
| XM_006256836.2 | XP_006256898.2 | 6 | 902 | |||
| XM_006256839.2 | XP_006256901.2 | 6 | 899 | |||
| XM_008773169.1 | XP_008771391.1 | 6 | 902 | |||
| GenBank mRNA | BC086548.1 | - | 5 | 513 | 7301795 | |
| Gene Symbol: LOC102547271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006256847.2 | XP_006256909.1 | 2 | 225 | ||

Msl3
Genomic Map