siRNA Details
| Gene Symbol: Zmynd12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014900.2 | NP_001014900.2 | 7 | 1064 | ||
| GenBank mRNA | AY959941.1 | - | 7 | 875 | ||
| BC147552.1 | - | 7 | 974 | 9056543 | ||
| BC147553.1 | - | 7 | 974 | 9056544 | ||
| BC147848.1 | - | 7 | 1064 | 9087565 | ||
| BC147851.1 | - | 7 | 1064 | 9087568 | ||

Zmynd12
Genomic Map