siRNA Details
| Gene Symbol: Tgif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015020.1 | NP_001015020.1 | 3 | 1053 | ||
| GenBank mRNA | BC091264.1 | - | 3 | 1053 | 7379061 | |
| BC127461.1 | - | 3 | 1141 | 7389058 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tgif1| Gene Symbol: Tgif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015020.1 | NP_001015020.1 | 3 | 1053 | ||
| GenBank mRNA | BC091264.1 | - | 3 | 1053 | 7379061 | |
| BC127461.1 | - | 3 | 1141 | 7389058 | ||