siRNA Details
| Gene Symbol: Gm5431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024230.2 | NP_001019401.2 | 5 | 2128 | ||
| GenBank mRNA | AK144287.1 | - | 5 | 2129 | ||
| AK165745.1 | - | 5 | 2129 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5431| Gene Symbol: Gm5431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024230.2 | NP_001019401.2 | 5 | 2128 | ||
| GenBank mRNA | AK144287.1 | - | 5 | 2129 | ||
| AK165745.1 | - | 5 | 2129 | |||