siRNA Details
| Gene Symbol: MGC109340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024267.2 | NP_001019438.1 | 1 | 1330 | ||
| GenBank mRNA | BC092634.1 | - | 1 | 1332 | 7315105 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGC109340| Gene Symbol: MGC109340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024267.2 | NP_001019438.1 | 1 | 1330 | ||
| GenBank mRNA | BC092634.1 | - | 1 | 1332 | 7315105 | |