siRNA Details
| Gene Symbol: FERMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017671.4 | NP_060141.3 | 4 | 1268 | ||
| GenBank mRNA | AB105105.1 | - | 4 | 1268 | ||
| AF443278.1 | - | 4 | 837 | |||
| AK092195.1 | - | 2 | 326 | |||
| AY137240.1 | - | 3 | 479 | |||
| BC035882.1 | - | 4 | 951 | 4941567 | ||
| BC040545.1 | - | 4 | 737 | 5277060 | ||

FERMT1
Genomic Map