siRNA Details
| Gene Symbol: Plac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024894.1 | NP_001020065.1 | 356 | |||
| GenBank mRNA | BC097979.1 | - | 356 | 7443160 | ||
| FQ219898.1 | - | 1 | 48 | |||
| FQ228726.1 | - | 649 | ||||
| Gene Symbol: LOC102550080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257666.2 | XP_006257728.1 | 368 | |||
| XM_006257669.2 | XP_006257731.1 | 3 | 350 | |||

Plac1
Genomic Map