siRNA Details
| Gene Symbol: MGC114492 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024895.1 | NP_001020066.1 | 4 | 1252 | ||
| XM_008773566.1 | XP_008771788.1 | 2 | 1237 | |||
| GenBank mRNA | BC097440.1 | - | 4 | 1252 | 7455831 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGC114492| Gene Symbol: MGC114492 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024895.1 | NP_001020066.1 | 4 | 1252 | ||
| XM_008773566.1 | XP_008771788.1 | 2 | 1237 | |||
| GenBank mRNA | BC097440.1 | - | 4 | 1252 | 7455831 | |