siRNA Details
| Gene Symbol: Pitpnm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024927.2 | NP_001020098.1 | 20 | 4365 | ||
| NM_001081641.1 | NP_001075110.1 | 19 | 4317 | |||
| GenBank mRNA | AK147444.1 | - | 20 | 4322 | ||
| AK147630.1 | - | 19 | 4150 | |||

Pitpnm3
Genomic Map