siRNA Details
| Gene Symbol: Gin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025012.1 | NP_001020183.1 | 8 | 1685 | ||
| XM_006245604.1 | XP_006245666.1 | 7 | 1608 | |||
| XM_006245605.2 | XP_006245667.1 | 8 | 2104 | |||
| XM_008767377.1 | XP_008765599.1 | 8 | 5280 | |||
| XM_008767378.1 | XP_008765600.1 | 8 | 4940 | |||
| GenBank mRNA | BC082058.1 | - | 8 | 1685 | 7132420 | |

Gin1
Genomic Map