siRNA Details
Gene Symbol: LOC547349 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025208.1 | NP_001020379.1 | 52 | |||
GenBank mRNA | AY533669.1 | - | 25 | |||
AY989869.1 | - | 52 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC547349 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025208.1 | NP_001020379.1 | 52 | |||
GenBank mRNA | AY533669.1 | - | 25 | |||
AY989869.1 | - | 52 |