siRNA Details
| Gene Symbol: LOC547349 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025208.1 | NP_001020379.1 | 1 | 278 | ||
| GenBank mRNA | AY533669.1 | - | 1 | 251 | ||
| AY989869.1 | - | 1 | 278 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC547349| Gene Symbol: LOC547349 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025208.1 | NP_001020379.1 | 1 | 278 | ||
| GenBank mRNA | AY533669.1 | - | 1 | 251 | ||
| AY989869.1 | - | 1 | 278 | |||