siRNA Details
| Gene Symbol: KPRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025231.1 | NP_001020402.1 | 2 | 1940 | ||
| XM_011509571.1 | XP_011507873.1 | 2 | 1981 | |||
| GenBank mRNA | AY960854.1 | - | 1 | 1911 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KPRP| Gene Symbol: KPRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025231.1 | NP_001020402.1 | 2 | 1940 | ||
| XM_011509571.1 | XP_011507873.1 | 2 | 1981 | |||
| GenBank mRNA | AY960854.1 | - | 1 | 1911 | ||