siRNA Details
| Gene Symbol: Gm20747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025241.3 | NP_001020412.1 | 1 | 12 | ||
| GenBank mRNA | BC089498.1 | - | 1 | 12 | 6744023 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm20747| Gene Symbol: Gm20747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025241.3 | NP_001020412.1 | 1 | 12 | ||
| GenBank mRNA | BC089498.1 | - | 1 | 12 | 6744023 | |