siRNA Details
Gene Symbol: Krtap31-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025244.3 | NP_001020415.1 | 1 | 786 | ||
GenBank mRNA | AK037252.1 | - | 1 | 789 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Krtap31-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025244.3 | NP_001020415.1 | 1 | 786 | ||
GenBank mRNA | AK037252.1 | - | 1 | 789 |