siRNA Details
| Gene Symbol: CXorf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025265.2 | NP_001020436.1 | 3 | 404 | ||
| NR_033212.1 | 3 | 385 | ||||
| XM_005262244.3 | XP_005262301.1 | 3 | 589 | |||
| GenBank mRNA | BC144434.1 | - | 3 | 202 | ||
| DB457782.1 | - | 3 | 360 | |||
| DB523273.1 | - | 2 | 88 | |||

CXorf65
Genomic Map