siRNA Details
| Gene Symbol: C3orf70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025266.1 | NP_001020437.1 | 2 | 5330 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C3orf70| Gene Symbol: C3orf70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025266.1 | NP_001020437.1 | 2 | 5330 | ||