siRNA Details
| Gene Symbol: Pnisr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025274.2 | NP_001020445.2 | 11 | 1597 | ||
| GenBank mRNA | AY283238.1 | - | 1 | 170 | ||
| BC091301.1 | - | 2 | 676 | 7309546 | ||
| FQ231658.1 | - | 4 | 444 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pnisr| Gene Symbol: Pnisr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025274.2 | NP_001020445.2 | 11 | 1597 | ||
| GenBank mRNA | AY283238.1 | - | 1 | 170 | ||
| BC091301.1 | - | 2 | 676 | 7309546 | ||
| FQ231658.1 | - | 4 | 444 | |||